N-Hydroxyurethane ≥85.0% (by GC, total nitrogen)
Supplier: TCI America
H0618-1G
H0618-5G
TCH0618-001GEA
32
USD
TCH0618-001G
TCH0618-5G
N-Hydroxyurethane ≥85.0% (by GC, total nitrogen)
N-Hydroxyurethane
CAS Number: 589-41-3
MDL Number: MFCD00002108
Molecular Formula: C3H7NO3
Molecular Weight: 105.09
Purity/Analysis Method: >85.0% (GC,N)
Form: Clear Liquid
Boiling point (°C): 113
Flash Point (°C): 110
Other Product Names / Synonyms: Ethyl Hydroxycarbamate, Hydroxycarbamic Acid Ethyl Ester
Formula:
C₃H₇NO₃ Boiling Pt: 113 °C Flash Pt: 110 °C |
MDL Number:
MFCD00002108 CAS Number: 589-41-3 |
Specification Test Results
Purity/Analysis Method | >85.0% (GC,N) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...