Ishikawa's reagent ≥92.0%

Supplier: TCI America

Synonyms: 1-(Diethylamino)-1,1,2,3,3,3-hexafluoropropane, Hexafluoropropene diethylamine, N,N-Diethyl-1,1,2,3,3,3-hexafluoropropylamine

H0598-25G H0598-100G
TCH0598-025GEA 79 USD
TCH0598-025G TCH0598-100G
Ishikawa's reagent ≥92.0%
Ishikawa's reagent

[Fluorinating Reagent]
CAS Number: 309-88-6
MDL Number: MFCD00060264
Molecular Formula: C7H11F6N
Molecular Weight: 223.16
Purity/Analysis Method: >92.0% (T)
Form: Clear Liquid
Boiling point (°C): 54
Flash Point (°C): 40
Specific Gravity (20/20): 1.23


Product Categories: Synthetic Organic Chemistry, Fluorinating Agents
Other Product Names / Synonyms: Hexafluoropropene Diethylamine, N,N-Diethyl-1,1,2,3,3,3-hexafluoropropylamine

Formula: C₇H₁₁F₆N
Boiling Pt: 54 °C
Density: 1.23 (20 °C)
Flash Pt: 40 °C
MDL Number: MFCD00060264
CAS Number: 309-88-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9a5186904a774484d51d5394744d38a6a97aa4e&vid=a9a5186904a774484d51d5394744d38a6a97aa4e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >92.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR