3-Hydroxypropanesulfonic acid ∼80% ∼7.8 mol/L in water (contains varying amounts of 3,3'-Oxydipropanesulfonic Acid)
Supplier: TCI America
H0597-500G
H0597-25G
TCH0597-500GEA
343
USD
TCH0597-500G
TCH0597-025G
3-Hydroxypropanesulfonic acid ∼80% ∼7.8 mol/L in water (contains varying amounts of 3,3'-Oxydipropanesulfonic Acid)
3-Hydroxypropanesulfonic acid
CAS Number: 15909-83-8
MDL Number: MFCD00066499
Molecular Formula: C3H8O4S
Molecular Weight: 140.15
Form: Clear Liquid
Color: Slightly Pale Yellow
Formula:
C₃H₈O₄S MW: 140.15 g/mol |
MDL Number:
MFCD00066499 CAS Number: 15909-83-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...