Hexadecanophenone ≥95.0%

Supplier: TCI America

H0590-25G H0590-5G
TCH0590-025GEA 165 USD
TCH0590-025G TCH0590-5G
Hexadecanophenone ≥95.0%
Hexadecanophenone

CAS Number: 6697-12-7
MDL Number: MFCD00008997
Molecular Formula: C22H36O
Molecular Weight: 316.53
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Boiling point (°C): 248
Melting point (°C): 59


Other Product Names / Synonyms: Palmitophenone, Pentadecyl Phenyl Ketone

Formula: C₂₂H₃₆O
Boiling Pt: 248 °C
Melting Pt: 59 °C
MDL Number: MFCD00008997
CAS Number: 6697-12-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=64dfac785798269064ad2d766a4bba03b4b1d55d&vid=64dfac785798269064ad2d766a4bba03b4b1d55d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR