DL-4-Hydroxyphenyllactic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 2-Hydroxy-3-(4-hydroxyphenyl)propanoic acid, Hydroxyphenyllactic acid, 2-Hydroxy-3-(4-hydroxyphenyl)propionic Acid

H0542-1G H0542-100MG
TCH0542-1GEA 517 USD
TCH0542-1G TCH0542-100MG
DL-4-Hydroxyphenyllactic acid ≥98.0% (by HPLC, titration analysis)
DL-4-Hydroxyphenyllactic acid

CAS Number: 306-23-0
MDL Number: MFCD00010514
Molecular Formula: C9H10O4
Molecular Weight: 182.18
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal


Other Product Names / Synonyms: 2-Hydroxy-3-(4-hydroxyphenyl)propionic Acid

Formula: C₉H₁₀O₄
MW: 182.18 g/mol
MDL Number: MFCD00010514
CAS Number: 306-23-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea0a92841b2a128d7ab61a17d8757b392a793c97&vid=ea0a92841b2a128d7ab61a17d8757b392a793c97&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR