4-Hydroxy-3-methoxyphenylpyruvic acid ≥95.0% (by HPLC)

Supplier: TCI America

Synonyms: Vanilpyruvic acid, 3-(4-Hydroxy-3-methoxyphenyl)-2-oxopropanoic acid

H0539-1G H0539-100MG
TCH0539-1GEA 571 USD
TCH0539-1G TCH0539-100MG
4-Hydroxy-3-methoxyphenylpyruvic acid ≥95.0% (by HPLC)
4-Hydroxy-3-methoxyphenylpyruvic acid

CAS Number: 1081-71-6
MDL Number: MFCD00056707
Molecular Formula: C10H10O5
Molecular Weight: 210.19
Purity/Analysis Method: >95.0% (HPLC)
Form: Crystal

Formula: C₁₀H₁₀O₅
MW: 210.19 g/mol
MDL Number: MFCD00056707
CAS Number: 1081-71-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2faee7ed78f0c50b9740522674dbc646b21370f6&vid=2faee7ed78f0c50b9740522674dbc646b21370f6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR