Hexamethonium bromide ≥98.0%

Supplier: TCI America

Synonyms: N1,N1,N1,N6,N6,N6-hexamethylhexane-1,6-diaminium bromide, Hexane-1,6-bis(trimethylammonium Bromide), Hexamethylenebis(trimethylammonium Bromide)

H0481-25G H0481-500G
TCH0481-025GEA 60 USD
TCH0481-025G TCH0481-500G
Hexamethonium bromide ≥98.0%
Hexamethonium bromide

CAS Number: 55-97-0
MDL Number: MFCD00011787
Molecular Formula: C12H30Br2N2
Molecular Weight: 362.19
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal
Melting point (°C): 285


Other Product Names / Synonyms: Hexamethylenebis(trimethylammonium Bromide), Hexane-1,6-bis(trimethylammonium Bromide)

Formula: C₁₂H₃₀Br₂N₂
Melting Pt: 285 °C
MDL Number: MFCD00011787
CAS Number: 55-97-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=69c329ce18772c68e66d0cba6da230cf8c03d85b&vid=69c329ce18772c68e66d0cba6da230cf8c03d85b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (N,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR