N-(3-Hydroxypropyl)ethylenediamine ≥97.0%
Supplier: TCI America
Synonyms:
2-(3-Hydroxypropylamino)ethylamine, N-(2-Aminoethyl)propanolamine, 3-((2-Aminoethyl)amino)propan-1-ol
H0391-25G
H0391-5G
TCH0391-025GEA
729
USD
TCH0391-025G
TCH0391-005G
N-(3-Hydroxypropyl)ethylenediamine ≥97.0%
N-(3-Hydroxypropyl)ethylenediamine
CAS Number: 56344-32-2
MDL Number: MFCD00059848
Molecular Formula: C5H14N2O
Molecular Weight: 118.18
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Boiling point (°C): 149
Melting point (°C): 49
Other Product Names / Synonyms: N-(2-Aminoethyl)propanolamine, 2-(3-Hydroxypropylamino)ethylamine
Formula:
C₅H₁₄N₂O MW: 118.18 g/mol Boiling Pt: 149 °C Melting Pt: 49 °C |
MDL Number:
MFCD00059848 CAS Number: 56344-32-2 |
|
Specification Test Results
Purity/Analysis Method | >97.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...