Sodium-DL-2-hydroxybutyrate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
DL-2-Hydroxybutyric Acid Sodium Salt, Sodium 2-hydroxybutanoate
H0387-5G
H0387-25G
TCH0387-005GEA
128
USD
TCH0387-005G
TCH0387-025G
Sodium-DL-2-hydroxybutyrate ≥98.0% (by titrimetric analysis)
Sodium-DL-2-hydroxybutyrate
CAS Number: 5094-24-6
MDL Number: MFCD00004565
Molecular Formula: C4H8O3
Molecular Weight: 126.09
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Other Product Names / Synonyms: DL-2-Hydroxybutyric Acid Sodium Salt
Formula:
C₄H₈O₃ MW: 126.09 g/mol |
MDL Number:
MFCD00004565 CAS Number: 5094-24-6 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...