1-[N-(2-Hydroxyethyl)-4'-piperidyl]-3-(4'-piperidyl)propane ≥96.0% (by GC, titration analysis)

Supplier: TCI America
Warning

H0361-25G H0361-5G
TCH0361-025GEA 388 USD
TCH0361-025G TCH0361-5G
1-[N-(2-Hydroxyethyl)-4'-piperidyl]-3-(4'-piperidyl)propane ≥96.0% (by GC, titration analysis)
1-[N-(2-Hydroxyethyl)-4'-piperidyl]-3-(4'-piperidyl)propane

CAS Number: 14712-23-3
MDL Number: MFCD00023766
Molecular Formula: C15H30N2O
Molecular Weight: 254.42
Purity/Analysis Method: >96.0% (GC,T)
Form: Crystal
Boiling point (°C): 250
Melting point (°C): 83

Formula: C₁₅H₃₀N₂O
MW: 254.42 g/mol
Boiling Pt: 250 °C
Melting Pt: 83 °C
MDL Number: MFCD00023766
CAS Number: 14712-23-3
EINECS: 238-761-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d1ab51bafeb65752eb7fdfde15b40693631f88cf&vid=2a126d8198e36e60bba8075313295a26f2c19213&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %
Purity (Nonaqueous Titration) min. 96.0 %
Melting point 82.0 to 86.0 deg-C
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR