Acetyl-L-carnitine hydrochloride ≥98.0%

Supplier: TCI America

Synonyms: (R)-3-Acetoxy-4-(trimethylammonio)butyrate Hydrochloride, (R)-2-Acetoxy-3-carboxy-N,N,N-trimethylpropan-1-aminium chloride, Acetyl-L-carnitine HCl

A1394-25G A1394-5G
TCA1394-25GEA 260.54 USD
TCA1394-25G TCA1394-005G
Acetyl-L-carnitine hydrochloride ≥98.0%
Acetyl-L-carnitine hydrochloride

CAS Number: 5080-50-2
MDL Number: MFCD00082230
Molecular Formula: C9H17NO4
Molecular Weight: 239.70
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Specific rotation [a]20/D: -28 deg (C=1, H2O)


Product Categories: Bioscience, Vitamin Related Compounds
Other Product Names / Synonyms: (R)-3-Acetoxy-4-(trimethylammonio)butyrate Hydrochloride

Formula: C₉H₁₇NO₄
MW: 239.70 g/mol
MDL Number: MFCD00082230
CAS Number: 5080-50-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c34b6b30078a03e9602b157e3199c802f8afd1cd&vid=c34b6b30078a03e9602b157e3199c802f8afd1cd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR