1,4-Dicyano-2-butene ≥98.0%
Supplier: TCI America
Synonyms:
Dihydromucononitrile
H0357-5G
H0357-25G
TCH0357-005GEA
56.3
USD
TCH0357-005G
TCH0357-025G
1,4-Dicyano-2-butene ≥98.0%
1,4-Dicyano-2-butene
CAS Number: 1119-85-3
MDL Number: MFCD00001964
Molecular Formula: C6H6N2
Molecular Weight: 106.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 120
Melting point (°C): 76
Other Product Names / Synonyms: 1,4-Dicyano-2-butene
Formula:
C₆H₆N₂ MW: 106.1252 g/mol Boiling Pt: 120 °C Melting Pt: 76 °C |
MDL Number:
MFCD00001964 CAS Number: 1119-85-3 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...