Heptanoic anhydride ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Enanthic Anhydride

H0353-25ML H0353-500ML
TCH0353-025MLEA 35 USD
TCH0353-025ML TCH0353-500ML
Heptanoic anhydride ≥97.0% (by titrimetric analysis)
Heptanoic anhydride

CAS Number: 626-27-7
MDL Number: MFCD00009536
Molecular Formula: C14H26O3
Molecular Weight: 242.36
Purity/Analysis Method: >97.0% (T)
Form: Clear Liquid
Boiling point (°C): 260
Melting point (°C): -12
Flash Point (°C): 110
Specific Gravity (20/20): 0.92


Other Product Names / Synonyms: Enanthic Anhydride

Formula: C₁₄H₂₆O₃
MW: 242.36 g/mol
Boiling Pt: 260 °C
Melting Pt: –12 °C
Density: 0.92 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00009536
CAS Number: 626-27-7

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR