2,4-Hexadiene ≥95.0%

Supplier: TCI America

Synonyms: Hexa-2,4-diene

H0336-5ML H0336-25ML
TCH0336-005MLEA 118.38 USD
TCH0336-005ML TCH0336-25ML
2,4-Hexadiene ≥95.0%
2,4-Hexadiene

CAS Number: 592-46-1
MDL Number: MFCD00009297
Molecular Formula: C6H10
Molecular Weight: 82.15
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 83
Flash Point (°C): -8
Specific Gravity (20/20): 0.72

Formula: C₆H₁₀
MW: 82.15 g/mol
Boiling Pt: 83 °C
Melting Pt: –79 °C
Density: 0.72 (20 °C)
Flash Pt: –8 °C
MDL Number: MFCD00009297
CAS Number: 592-46-1
EINECS: 209-757-3
UN: 2458
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3511f435fcd3f71f8178f2e737208975aef6dab0&vid=59a53fafffb63763d4ac2c8759590aa15546aa81&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR