3-Hydroxy-2'-methoxy-2-naphthanilide ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
CAS Number: 135-62-6
MDL Number: MFCD00021630
Molecular Formula: C18H15NO3
Molecular Weight: 293.32
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 162
Other Product Names / Synonyms: 2-Hydroxy-3-naphthoic Acid o-Anisidide, Naphthol AS-OL, 3-Hydroxy-N-(2-methoxyphenyl)-2-naphthamide, Azoic Coupling Component 20
Formula:
C₁₈H₁₅NO₃ Melting Pt: 162 °C |
MDL Number:
MFCD00021630 CAS Number: 135-62-6 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...