Naphthol AS-D ≥97.0% (by HPLC)
Supplier: TCI America
Synonyms:
3-Hydroxy-2'-methyl-2-naphthanilide, NSC-37188, Azoic Coupling Component 18, Naphthol AS-D, 3-Hydroxy-N-(o-tolyl)-2-naphthamide, 2-Hydroxy-3-naphthoic acid o-Toluidide
H0315-25G
H0315-500G
TCH0315-025GEA
72
USD
TCH0315-025G
TCH0315-500G
Naphthol AS-D ≥97.0% (by HPLC)
Naphthol AS-D
CAS Number: 135-61-5
MDL Number: MFCD00021634
Molecular Formula: C18H15NO2
Molecular Weight: 277.32
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Melting point (°C): 198
Other Product Names / Synonyms: 2-Hydroxy-3-naphthoic Acid o-Toluidide, Naphthol AS-D, Azoic Coupling Component 18, 3-Hydroxy-N-(o-tolyl)-2-naphthamide
Formula:
C₁₈H₁₅NO₂ MW: 277.32 g/mol Melting Pt: 198 °C |
MDL Number:
MFCD00021634 CAS Number: 135-61-5 EINECS: 205-205-0 |
Specification Test Results
Purity/Analysis Method | >97.0% (HPLC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...