3-Hydroxypropionic acid 30% ∼3.6 mol/L in water (contains varying amounts of 3,3'-Oxydipropionic Acid) for synthesis
Supplier: TCI America
Synonyms:
3-Hydroxypropionic Acid, 3-Hydroxypropionic Acid (contains varying amounts of 3,3-Oxydipropionic Acid), Hydracrylic Acid
H0297-25G
H0297-10G
TCH0297-25GEA
386
USD
TCH0297-25G
TCH0297-010G
3-Hydroxypropionic acid 30% ∼3.6 mol/L in water (contains varying amounts of 3,3'-Oxydipropionic Acid) for synthesis
3-Hydroxypropionic acid
CAS Number: 503-66-2
Molecular Formula: C3H6O3
Molecular Weight: 90.08
Form: Liquid
Color: Colorless
Formula:
C₃H₆O₃ MW: 90.08 g/mol |
MDL Number:
MFCD00058998 CAS Number: 503-66-2 |
Specification Test Results
Specific Gravity (20/20) | 1.08 |
Storage Temperature | 0-10°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...