5-Hydroxy-1,4-naphthoquinone (juglone) ≥97.0%
Supplier: TCI America
Synonyms:
5-Hydroxy-p-naphthoquinone, Juglone, 5-Hydroxy-1,4-naphthalenedione
H0286-5G
H0286-1G
TCH0286-005GEA
359.9
USD
TCH0286-005G
TCH0286-001G
5-Hydroxy-1,4-naphthoquinone (juglone) ≥97.0%
5-Hydroxy-1,4-naphthoquinone (juglone)
CAS Number: 481-39-0
MDL Number: MFCD00001684
Molecular Formula: C10H6O3
Molecular Weight: 174.16
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 155
Other Product Names / Synonyms: Juglone
Formula:
C₁₀H₆O₃ MW: 174.16 g/mol Melting Pt: 155 °C |
MDL Number:
MFCD00001684 CAS Number: 481-39-0 UN: 2811 ADR: 6.1 III |
|
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...