4-Hydroxysophthalic acid ≥98.0% for synthesis

Supplier: TCI America
Warning

Synonyms: 4-Hydroxyisophthalic acid

H0257-25G H0257-5G
TCH0257-025GEA 274 USD
TCH0257-025G TCH0257-5G
4-Hydroxysophthalic acid ≥98.0% for synthesis
4-Hydroxysophthalic acid

CAS Number: 636-46-4
MDL Number: MFCD00010391
Molecular Formula: C8H6O5
Molecular Weight: 182.13
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal

Formula: C₈H₆O₅
Melting Pt: 316 °C
MDL Number: MFCD00010391
CAS Number: 636-46-4
EINECS: 211-258-0
Merck Index: 12,04873
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=479bf5a200c9c9e95620e44266f4ced847217872&vid=b06b14ee1f9c9580d3e7afc0e7ac13bd587324f5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR