Methyl-2-hydroxyisobutyrate ≥99.5%

Supplier: TCI America

Synonyms: Methyl 2-Hydroxyisobutyrate, 2-Hydroxyisobutyric Acid Methyl Ester

H0256-25ML H0256-500ML H0256-100ML
TCH0256-025MLEA 41 USD
TCH0256-025ML TCH0256-500ML TCH0256-100ML
Methyl-2-hydroxyisobutyrate ≥99.5%
Methyl-2-hydroxyisobutyrate

CAS Number: 2110-78-3
MDL Number: MFCD00004457
Molecular Formula: C5H10O3
Molecular Weight: 118.13
Purity/Analysis Method: >99.5% (GC)
Form: Clear Liquid
Boiling point (°C): 137
Flash Point (°C): 45
Specific Gravity (20/20): 1.03


Other Product Names / Synonyms: 2-Hydroxyisobutyric Acid Methyl Ester

Formula: C₅H₁₀O₃
MW: 118.13 g/mol
Boiling Pt: 137 °C
Density: 1.03 (20 °C)
Flash Pt: 45 °C
MDL Number: MFCD00004457
CAS Number: 2110-78-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=75b45f86be1400988b8de9134bee5052991368f7&vid=75b45f86be1400988b8de9134bee5052991368f7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.5% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR