Hydrazinium monochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

Synonyms: Hydrazinium monochloride, Hydrazinium chloride, Hydrazine monohydrochloride, Hydrazine hydrochloride

H0174-25G H0174-500G
TCH0174-025GEA 47.83 USD
TCH0174-025G TCH0174-500G
Hydrazinium monochloride ≥98.0% (by titrimetric analysis)
Hydrazinium monochloride

CAS Number: 2644-70-4
MDL Number: MFCD00044368
Molecular Formula: H4N2
Molecular Weight: 68.50
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 93


Product Categories: Synthetic Organic Chemistry, Reduction
Other Product Names / Synonyms: Hydrazinium Monochloride

Formula: H₅ClN₂
MW: 68.51 g/mol
Melting Pt: 93 °C
Density: 1.5 g/cm³ (20 °C)
MDL Number: MFCD00044368
CAS Number: 2644-70-4
EINECS: 220-154-4
UN: 2923
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82b375cf97d909c2b11af609d7f797b84ffb3b0b&vid=82b375cf97d909c2b11af609d7f797b84ffb3b0b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR