Hydrazine dihydrobromide hydrate ≥98.0%
Supplier: TCI America
Synonyms:
Hydrazinium Dibromide
H0169-25G
H0169-500G
TCH0169-025GEA
50
USD
TCH0169-025G
TCH0169-500G
Hydrazine dihydrobromide hydrate ≥98.0%
Hydrazine dihydrobromide hydrate
CAS Number: 23268-00-0
MDL Number: MFCD00070601
Molecular Formula: H4N2
Molecular Weight: 193.87
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Boiling point (°C): 178
Flash Point (°C): 191
Product Categories: Synthetic Organic Chemistry, Reduction
Other Product Names / Synonyms: Hydrazinium Dibromide
Formula:
H₄N₂ Boiling Pt: 178 °C Flash Pt: 191 °C |
MDL Number:
MFCD00070601 CAS Number: 23268-00-0 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...