Histamine diphosphate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2-(4-Imidazolyl)ethylamine diphosphate salt, 2-(1H-Imidazol-4-yl)ethan-1-amine bis(phosphate), Histamine phosphate

H0147-1G H0147-5G
TCH0147-001GEA 45 USD
TCH0147-001G TCH0147-005G
Histamine diphosphate ≥98.0% (by titrimetric analysis)
Histamine diphosphate

CAS Number: 51-74-1
MDL Number: MFCD00064316
Molecular Formula: C5H9N3
Molecular Weight: 307.14
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 133
Storage Temperature: 0-10°C

Formula: C₅H₉N₃
MW: 307.14 g/mol
Melting Pt: 133 °C
MDL Number: MFCD00064316
CAS Number: 51-74-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6aa7fbbc7a22086e2f54d78eeb039b4446c7baa1&vid=0f48db86851ce26fcc708c74b45d8bcca193bd55&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR