Hexadecyltrimethylammonium hydroxide 25% in methanol

Supplier: TCI America

Synonyms: Cetyltrimethylammonium Hydroxide

H0083-500ML H0083-25ML
TCH0083-500MLEA 325.81 USD
TCH0083-500ML TCH0083-025ML
Hexadecyltrimethylammonium hydroxide 25% in methanol
Hexadecyltrimethylammonium hydroxide

CAS Number: 505-86-2
MDL Number: MFCD00041921
Molecular Formula: C19H43NO
Molecular Weight: 301.56
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 11
Specific Gravity (20/20): 0.83


Other Product Names / Synonyms: Cetyltrimethylammonium Hydroxide

Formula: C₁₉H₄₃NO
MW: 301.56 g/mol
Density: 0.83 (20 °C)
Flash Pt: 11 °C
MDL Number: MFCD00041921
CAS Number: 505-86-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e521f5c70194cef241b9d9a4166239e98dd325d7&vid=fb63b02f73dfbf32df9b0c63c4b5c12f67ef877d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR