Hexadecanamide ≥95.0%

Supplier: TCI America

H0067-10G H0067-25G
TCH0067-010GEA 198.64 USD
TCH0067-010G TCH0067-025G
Hexadecanamide ≥95.0%
Hexadecanamide

CAS Number: 629-54-9
MDL Number: MFCD00025534
Molecular Formula: C16H33NO
Molecular Weight: 255.45
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Boiling point (°C): 236
Melting point (°C): 106


Product Categories: Materials Chemistry, Pressure & Heat Sensitive Recording Materials
Other Product Names / Synonyms: Palmitamide

Formula: C₁₆H₃₃NO
Boiling Pt: 236 °C
Melting Pt: 106 °C
MDL Number: MFCD00025534
CAS Number: 629-54-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16c3479b814ac48ca94988b2822cf5bd7409bf7b&vid=16c3479b814ac48ca94988b2822cf5bd7409bf7b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR