(±)-3-Heptanol ≥98.0%

Supplier: TCI America

Synonyms: 3-Heptyl alcohol, 3-Heptanol, Heptan-3-ol, Butylethylcarbinol, (±)-Ethyl butyl carbinol, (±)-Butyl ethyl carbinol, (±)-3-Hydroxy heptane

H0035-25ML H0035-500ML
TCH0035-025MLEA 69.2 USD
TCH0035-025ML TCH0035-500ML
(±)-3-Heptanol ≥98.0%
(±)-3-Heptanol

CAS Number: 589-82-2
MDL Number: MFCD00004586
Molecular Formula: C7H16O
Molecular Weight: 116.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 156
Flash Point (°C): 60
Specific Gravity (20/20): 0.82


Other Product Names / Synonyms: Butylethylcarbinol, 3-Heptyl Alcohol

Formula: C₇H₁₆O
MW: 116.2 g/mol
Boiling Pt: 156 °C
Melting Pt: -70 °C
Density: 0.82 (20 °C)
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00004586
CAS Number: 589-82-2
EINECS: 209-661-1
UN: 1987
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=748a61bdc580a5cb329259f9747bc9a151618835&vid=f74db19b9c6da94b8e88755f773e99ec750be3b1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR