2-Heptadecylimidazole ≥85.0%

Supplier: TCI America

H0021-500G H0021-25G
TCH0021-500GEA 530.4 USD
TCH0021-500G TCH0021-025G
2-Heptadecylimidazole ≥85.0%
2-Heptadecylimidazole

CAS Number: 23328-87-2
MDL Number: MFCD00038620
Molecular Formula: C20H38N2
Molecular Weight: 306.54
Purity/Analysis Method: >85.0% (GC)
Form: Crystal
Melting point (°C): 89
Flash Point (°C): 245

Formula: C₂₀H₃₈N₂
Melting Pt: 89 °C
Flash Pt: 245 °C
MDL Number: MFCD00038620
CAS Number: 23328-87-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1468d151494ebe10131be0b9bd391c859261c573&vid=1468d151494ebe10131be0b9bd391c859261c573&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >85.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR