Geldanamycin ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: (4E,6Z,8S,9S,10E,12S,13R,14S,16R)-13-Hydroxy-8,14,19-trimethoxy-4,10,12,16-tetramethyl-3,20,22-trioxo-2-azabicyclo[16.3.1]docosa-1(21),4,6,10,18-pentaen-9-yl carbamate

G0334-10MG
TCG0334-10MGEA 386.72 USD
TCG0334-10MG
Geldanamycin ≥98.0% (by HPLC)
Geldanamycin

CAS Number: 30562-34-6
MDL Number: MFCD00274570
Molecular Formula: C29H40N2O9
Molecular Weight: 560.64
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Melting point (°C): 255
Lambda max.: 305 nm (MeOH)Storage Temperature: 0°C

Formula: C₂₉H₄₀N₂O₉
MW: 560.64 g/mol
Melting Pt: 255 °C
MDL Number: MFCD00274570
CAS Number: 30562-34-6
https://pr.vwr.com/stibo/web/std.lang.all/99/44/28969944.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR