4-Glycidyloxycarbazole ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: 4-(2,3-Epoxypropoxy)carbazole, Carvedilol Related Compound D

G0296-5G G0296-25G
TCG0296-5GEA 42 USD
TCG0296-5G TCG0296-25G
4-Glycidyloxycarbazole ≥98.0% (by HPLC, total nitrogen)
4-Glycidyloxycarbazole

CAS Number: 51997-51-4
MDL Number: MFCD03411884
Molecular Formula: C15H13NO2
Molecular Weight: 239.27
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 4-(2,3-Epoxypropoxy)carbazole

Formula: C₁₅H₁₃NO₂
MW: 239.27 g/mol
MDL Number: MFCD03411884
CAS Number: 51997-51-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f92d7c45d5736d4b720eccf319e70222558b796&vid=6f92d7c45d5736d4b720eccf319e70222558b796&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR