3-Methoxyanthranilic acid ≥99.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
2-Amino-m-anisic acid, 3-Methoxyanthranilic acid, o-Anisidine-6-carboxylic acid, 2-Amino-3-methoxybenzoic acid
A1378-25G
A1378-5G
TCA1378-25GEA
290
USD
TCA1378-25G
TCA1378-005G
3-Methoxyanthranilic acid ≥99.0% (by HPLC, titration analysis)
3-Methoxyanthranilic acid
CAS Number: 3177-80-8
MDL Number: MFCD00075178
Molecular Formula: C8H9NO3
Molecular Weight: 167.16
Purity/Analysis Method: >99.0% (HPLC,T)
Form: Crystal
Melting point (°C): 172
Product Categories: Synthetic Organic Chemistry, Benzyne Precursors (Aryne Precursors)
Other Product Names / Synonyms: 2-Amino-m-anisic Acid, o-Anisidine-6-carboxylic Acid, 3-Methoxyanthranilic Acid
Formula:
C₈H₉NO₃ MW: 167.16 g/mol Melting Pt: 172 °C Storage Temperature: Ambient |
MDL Number:
MFCD00075178 CAS Number: 3177-80-8 |
Specification Test Results
Purity/Analysis Method | >99.0% (HPLC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...