N-(tert-Butoxycarbonyl)-1,4-diaminobutane ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
N-(4-Aminobutyl)carbamic Acid tert-Butyl Ester, N-(tert-Butoxycarbonyl)-1,4-butanediamine, NH2-C4-NH-Boc, N-Boc-1,4-diaminobutane, N-Boc-1,4-butanediamine, tert-Butyl N-(4-Aminobutyl)carbamate, tert-Butyl (4-aminobutyl)carbamate
CAS Number: 68076-36-8
MDL Number: MFCD00210019
Molecular Formula: C9H20N2O2
Molecular Weight: 188.27
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Specific Gravity (20/20): 0.99
Product Categories: Bioscience, Chemical Building Blocks for Polyamines
Other Product Names / Synonyms: N-(4-Aminobutyl)carbamic Acid tert-Butyl Ester, N-Boc-1,4-butanediamine, N-Boc-1,4-diaminobutane, N-(tert-Butoxycarbonyl)-1,4-butanediamine, tert-Butyl N-(4-Aminobutyl)carbamate
Formula:
C₉H₂₀N₂O₂ MW: 188.27 g/mol Density: 0.99 (20 °C) |
MDL Number:
MFCD00210019 CAS Number: 68076-36-8 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...