Glycidyl isopropyl ether ≥96.0%

Supplier: TCI America

Synonyms: 2,3-Epoxypropyl Isopropyl Ether

G0255-500ML G0255-25ML
TCG0255-500MLEA 161.38 USD
TCG0255-500ML TCG0255-025ML
Glycidyl isopropyl ether ≥96.0%
Glycidyl isopropyl ether

CAS Number: 4016-14-2
MDL Number: MFCD00005139
Molecular Formula: C6H12O2
Molecular Weight: 116.16
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 132
Flash Point (°C): 33
Specific Gravity (20/20): 0.92


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 2,3-Epoxypropyl Isopropyl Ether

Formula: C₆H₁₂O₂
Boiling Pt: 132 °C
Density: 0.92 (20 °C)
Flash Pt: 33 °C
MDL Number: MFCD00005139
CAS Number: 4016-14-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a1b08933a7c1f3cbbf7eeda534afd3b11d278396&vid=a1b08933a7c1f3cbbf7eeda534afd3b11d278396&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR