Glycyrrhetic acid ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 18 beta-Glycyrrhetic acid

G0149-1G G0149-25G
TCG0149-001GEA 26.45 USD
TCG0149-001G TCG0149-025G
Glycyrrhetic acid ≥97.0% (by titrimetric analysis)
Glycyrrhetic acid

CAS Number: 471-53-4 MDL Number: MFCD00003706 Molecular Formula: C30H46O4 Molecular Weight: 470.69 Purity/Analysis Method: 97.0% (T) Form: Crystal Melting point (°C): 297 Specific rotation [a]20/D: 162 deg (C=1, MeOH)

Formula: C₃₀H₄₆O₄
MW: 470.69 g/mol
Melting Pt: 297 °C
MDL Number: MFCD00003706
CAS Number: 471-53-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=891108170432264bdae91c2603223355e103cac8&vid=891108170432264bdae91c2603223355e103cac8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR