Glycyl-L-Tryptophan Hydrate

Supplier: TCI America

G0144-1G G0144-100MG
TCG0144-001GEA 81 USD
TCG0144-001G TCG0144-100MG
Glycyl-L-Tryptophan Hydrate
Proteins and Peptides

Product Categories: Bioscience, Oligopeptides
Other Product Names / Synonyms: H-Gly-Trp-OH


  • Specific rotation [a]20/D: 27.0 to 30.0° (C=2, 5 mol/L HCl)
  • Solubility in 5 mol/L HCl: almost transparency
  • EINECS: 219-235-7


Product Categories: Bioscience, Oligopeptides Other Product Names / Synonyms: H-Gly-Trp-OH

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=31c0be7b25ef385966b426d09b7c590676ab5c6c&vid=31c0be7b25ef385966b426d09b7c590676ab5c6c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR