Calcium glycerophosphate ≥95.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Calcium 2,3-dihydroxypropyl phosphate, Glycerophosphoric Acid Calcium Salt

G0095-500G G0095-25G
TCG0095-500GEA 150 USD
TCG0095-500G TCG0095-025G
Calcium glycerophosphate ≥95.0% (by titrimetric analysis)
Calcium glycerophosphate

CAS Number: 27214-00-2
MDL Number: MFCD00042643
Molecular Formula: C3H9O6P
Molecular Weight: 210.13
Purity/Analysis Method: >95.0% (T)
Form: Crystal


Other Product Names / Synonyms: Glycerophosphoric Acid Calcium Salt

Formula: C₃H₇CaO₆P
MW: 210.13 g/mol
MDL Number: MFCD00042643
CAS Number: 27214-00-2

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR