D-Glucose-6-phosphate barium salt heptahydrate ≥98.0%

Supplier: TCI America

Synonyms: D-Glucose-6-Phosphate Barium Salt Hydrate

G0052-100MG G0052-1G
TCG0052-100MGEA 24 USD
TCG0052-100MG TCG0052-001G
D-Glucose-6-phosphate barium salt heptahydrate ≥98.0%
D-Glucose-6-phosphate barium salt heptahydrate

CAS Number: 58823-95-3
MDL Number: MFCD00221521
Molecular Formula: C6H13O9P
Molecular Weight: 395.45
Purity/Analysis Method: >98.0% (W)
Form: Crystal
Specific rotation [a]20/D: 20 deg (C=5, 1mol/L HCl)
Storage Temperature: 0-10°C

Formula: C₆H₁₃O₉P
MDL Number: MFCD00221521
CAS Number: 58823-95-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d5e72a35eb35a40529f8e5c3564b344deb2d8a0&vid=6d5e72a35eb35a40529f8e5c3564b344deb2d8a0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (W)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR