β-D-Glucose ≥85.0% (contains alpha-D-Glucose)

Supplier: TCI America

G0047-25G G0047-500G
TCG0047-025GEA 63.76 USD
TCG0047-025G TCG0047-500G
β-D-Glucose ≥85.0% (contains alpha-D-Glucose)
β-D-Glucose

CAS Number: 492-61-5
MDL Number: MFCD00063989
Molecular Formula: C6H12O6
Molecular Weight: 180.16
Purity/Analysis Method: >85.0% (GC)
Form: Crystal
Melting point (°C): 155
Specific rotation [a]20/D: 53 deg (C=10, H2O)
Storage Temperature: 0-10°C

Formula: C₆H₁₂O₆
Melting Pt: 155 °C
MDL Number: MFCD00063989
CAS Number: 492-61-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=35e8fe6e7ae066899a84448634d055c1c6165d9c&vid=35e8fe6e7ae066899a84448634d055c1c6165d9c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >85.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR