4-Fluoro-2,1,3-benzoxadiazole ≥98.0%

Supplier: TCI America

Synonyms: 4-Fluorobenzofurazan

F0702-1G F0702-5G
TCF0702-1GEA 181 USD
TCF0702-1G TCF0702-5G
4-Fluoro-2,1,3-benzoxadiazole ≥98.0%
4-Fluoro-2,1,3-benzoxadiazole

CAS Number: 29270-55-1
MDL Number: MFCD03425566
Molecular Formula: C6H3FN2O
Molecular Weight: 138.10
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 83
Specific Gravity (20/20): 1.34


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Fluorobenzofurazan

Formula: C₆H₃FN₂O
MW: 138.10 g/mol
Boiling Pt: 83 °C
Density: 1.34 (20 °C)
MDL Number: MFCD03425566
CAS Number: 29270-55-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=adf666f88918339c7cbe35b8720094a4f7a47e78&vid=adf666f88918339c7cbe35b8720094a4f7a47e78&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR