Fexofenadine hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
2-(4-(1-Hydroxy-4-(4-(hydroxydiphenylmethyl)piperidin-1-yl)butyl)phenyl)-2-methylpropanoic acid hydrochloride, Fexofenadine HCl
F0698-1G
F0698-5G
TCF0698-1GEA
89.91
USD
TCF0698-1G
TCF0698-5G
Fexofenadine hydrochloride ≥98.0% (by HPLC, titration analysis)
Fexofenadine hydrochloride
CAS Number: 153439-40-8
MDL Number: MFCD00865710
Molecular Formula: C32H39NO4
Molecular Weight: 538.13
Purity/Analysis Method:
Formula:
C₃₂H₃₉NO₄ MW: 538.13 g/mol |
MDL Number:
MFCD00865710 CAS Number: 153439-40-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...