Fluconazole ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-(2,4-Difluorophenyl)-1,3-bis(1H-1,2,4-triazol-1-yl)propan-2-ol, 2-(2,4-Difluorophenyl)-1,3-di(1H-1,2,4-triazol-1-yl)propan-2-ol

F0677-1G F0677-5G
TCF0677-1GEA 87.9 USD
TCF0677-1G TCF0677-5G
Fluconazole ≥98.0% (by GC, titration analysis)
Fluconazole

CAS Number: 86386-73-4
Molecular Formula: C13H12F2N6O
Molecular Weight: 306.28
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 141


Product Categories: Synthetic Organic Chemistry, Fluorinated Bioactive Compounds, Bioscience, Antifungal Ingredients
Other Product Names / Synonyms: 2-(2,4-Difluorophenyl)-1,3-bis(1H-1,2,4-triazol-1-yl)propan-2-ol

Formula: C₁₃H₁₂F₂N₆O
MW: 306.28 g/mol
Melting Pt: 141 °C
MDL Number: MFCD00274549
CAS Number: 86386-73-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5298fa4ba79359113ee478a7cbd64c58c0f6d426&vid=5298fa4ba79359113ee478a7cbd64c58c0f6d426&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR