Fluocinolone acetonide ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 6alpha,9alpha-Difluoro-11beta,16alpha,17alpha,21-tetrahydroxy-1,4-pregnadiene-3,20-dione 16,17-Acetonide

F0657-1G F0657-5G
TCF0657-1GEA 202 USD
TCF0657-1G TCF0657-5G
Fluocinolone acetonide ≥98.0% (by HPLC)
Fluocinolone acetonide

CAS Number: 67-73-2
MDL Number: MFCD00010525
Molecular Formula: C24H30F2O6
Molecular Weight: 452.49
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 266
Specific rotation [a]20/D: 103 deg (C=1, MeOH)


Product Categories: Synthetic Organic Chemistry, Fluorinated Bioactive Compounds, Bioscience, Immune System, Inflammation, Allergy Research Reagents, Pregnanes (Glucocorticoids)
Other Product Names / Synonyms: 6alpha,9alpha-Difluoro-11beta,16alpha,17alpha,21-tetrahydroxy-1,4-pregnadiene-3,20-dione 16,17-Acetonide

Formula: C₂₄H₃₀F₂O₆
MW: 452.49 g/mol
Melting Pt: 266 °C
MDL Number: MFCD00010525
CAS Number: 67-73-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=97ff839939af451056234a2b3942e4e496ebd77f&vid=97ff839939af451056234a2b3942e4e496ebd77f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR