N-ɑ-Fmoc-N-Trityl-L-histidine ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: N-ɑ-Fmoc-tele-trityl-L-histidine, Fmoc-His(Trt)-OH, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(1-trityl-1H-imidazol-4-yl)propanoic acid, N-ɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-tele-(triphenylmethyl)-L-histidine, N-Fmoc-1-trityl-L-histidine

F0653-5G F0653-25G
TCF0653-5GEA 22 USD
TCF0653-5G TCF0653-25G
N-ɑ-Fmoc-N-Trityl-L-histidine ≥98.0% (by HPLC)
N-alpha-Fmoc-N-Trityl-L-histidine

CAS Number: 109425-51-6
MDL Number: MFCD00043332
Molecular Formula: C40H33N3O4
Molecular Weight: 619.72
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Specific rotation [a]20/D: 97 deg (C=5, CHCl3)


Product Categories: Synthetic Organic Chemistry, Peptide Synthesis, Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Nalpha-Fmoc-tele-trityl-L-histidine, Fmoc-His(Trt)-OH

Formula: C₄₀H₃₃N₃O₄
MW: 619.72 g/mol
MDL Number: MFCD00043332
CAS Number: 109425-51-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6da49df55ada716f038039a7a6c8dcf164186f92&vid=6da49df55ada716f038039a7a6c8dcf164186f92&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR