Fleroxacin ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Megalone, 6,8-Difluoro-1-(2-fluoroethyl)-1,4-dihydro-7-(4-methyl-1-piperazinyl)-4-oxo-3-quinolinecarboxylic Acid, 6,8-Difluoro-1-(2-fluoroethyl)-7-(4-methylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
CAS Number: 79660-72-3
MDL Number: MFCD00864880
Molecular Formula: C17H18F3N3O3
Molecular Weight: 369.34
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Product Categories: Synthetic Organic Chemistry, Fluorinated Bioactive Compounds, Bioscience, Synthetic Antimicrobial Ingredients
Other Product Names / Synonyms: 6,8-Difluoro-1-(2-fluoroethyl)-1,4-dihydro-7-(4-methyl-1-piperazinyl)-4-oxo-3-quinolinecarboxylic Acid, Megalone
Formula:
C₁₇H₁₈F₃N₃O₃ MW: 369.34 g/mol |
MDL Number:
MFCD00864880 CAS Number: 79660-72-3 |
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...