4-Fluorodiphenylamine ≥98.0%

Supplier: TCI America

Synonyms: 4-Fluoro-N-phenylaniline

F0630-1G F0630-5G
TCF0630-1GEA 105.7 USD
TCF0630-1G TCF0630-5G
4-Fluorodiphenylamine ≥98.0%
4-Fluorodiphenylamine

CAS Number: 330-83-6
MDL Number: MFCD01318538
Molecular Formula: C12H10FN
Molecular Weight: 187.22
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 166
Melting point (°C): 38
Flash Point (°C): 110
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Materials Chemistry, Building Blocks for Small Molecule Semiconductors
Other Product Names / Synonyms: N-(4-Fluorophenyl)aniline

Formula: C₁₂H₁₀FN
MW: 187.2129 g/mol
Boiling Pt: 166 °C
Melting Pt: 38 °C
Flash Pt: 110 °C
MDL Number: MFCD01318538
CAS Number: 330-83-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2faeae48eed027cdc28b63d0de7a09789b46c71&vid=e2faeae48eed027cdc28b63d0de7a09789b46c71&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR