4-Fluoropyridine hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
F0628-5G
F0628-1G
TCF0628-5GEA
339.86
USD
TCF0628-5G
TCF0628-1G
4-Fluoropyridine hydrochloride ≥98.0% (by HPLC, titration analysis)
4-Fluoropyridine hydrochloride
CAS Number: 39160-31-1
MDL Number: MFCD00234055
Molecular Formula: C5H4FN
Molecular Weight: 133.55
Purity/Analysis Method:
Formula:
C₅H₅ClFN MW: 133.55 g/mol Melting Pt: 100 °C |
MDL Number:
MFCD00234055 CAS Number: 39160-31-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...