Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-lysine hydrochloride ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: N2-Fmoc-D-lysine Hydrochloride, Fmoc-D-Lys-OH.HCl, Nɑ-Fmoc-D-lysine Hydrochloride

F0604-1G F0604-5G
TCF0604-1GEA 73 USD
TCF0604-1G TCF0604-5G
Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-lysine hydrochloride ≥98.0% (by HPLC)
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-lysine hydrochloride

CAS Number: 201002-47-3
MDL Number: MFCD00190888
Molecular Formula: C21H24N2O4
Molecular Weight: 404.89
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Specific rotation [a]20/D: 11 deg (C=1, DMF)
Storage Temperature: 0-10°C


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Nalpha-Fmoc-D-lysine Hydrochloride, Fmoc-D-Lys-OH.HCl

Formula: C₂₁H₂₄N₂O₄
MW: 404.89 g/mol
MDL Number: MFCD00190888
CAS Number: 201002-47-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=78356f662b37bb394728b35ceb5307a5acd1b43b&vid=78356f662b37bb394728b35ceb5307a5acd1b43b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR