2-Fluorobenzenesulfonamide ≥98.0%

Supplier: TCI America

F0557-25G F0557-5G
TCF0557-25GEA 226 USD
TCF0557-25G TCF0557-5G
2-Fluorobenzenesulfonamide ≥98.0%
2-Fluorobenzenesulphonamide

CAS Number: 30058-40-3
MDL Number: MFCD00042283
Molecular Formula: C6H6FNO2S
Molecular Weight: 175.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 161

Formula: C₆H₆FNO₂S
Melting Pt: 161 °C
MDL Number: MFCD00042283
CAS Number: 30058-40-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32ea990e5c240b3ecd8d902cedfceb505b0619c0&vid=32ea990e5c240b3ecd8d902cedfceb505b0619c0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR