N-(4-Fluorophenyl)formamide ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
4'-Fluoroformanilide, 1-Fluoro-4-formamidobenzene
F0546-5G
F0546-1G
TCF0546-5GEA
229
USD
TCF0546-5G
TCF0546-1G
N-(4-Fluorophenyl)formamide ≥98.0% (by GC)
N-(4-Fluorophenyl)formamide
CAS Number: 459-25-6
Molecular Formula: C7H6FNO
Molecular Weight: 139.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 67
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1-Fluoro-4-formamidobenzene, 4'-Fluoroformanilide
Formula:
C₇H₆FNO Melting Pt: 67 °C |
CAS Number:
459-25-6 |
|
Specification Test Results
Appearance | White to Gray to Red powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 66.0 to 70.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...