4-Fluorobenzhydrazide ≥98.0% (by GC, titration analysis)
Supplier: TCI America
CAS Number: 456-06-4
Molecular Formula: C7H7FN2O
Molecular Weight: 154.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 165
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Fluorobenzoic Hydrazide, 4-Fluorobenzoylhydrazine
Formula:
C₇H₇FN₂O Melting Pt: 165 °C |
MDL Number:
MFCD00060562 CAS Number: 456-06-4 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...