FMOC-O-tert-butyl-L-tyrosine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: N-α-Fmoc-o-t-butyl-L-Tyrosine, N-Fmoc-O-tert-butyl-L-tyrosine, Nalpha-Fmoc-O-tert-butyl-L-tyrosine, O-tert-Butyl-Nalpha-Fmoc-L-tyrosine, Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-O-tert-butyl-L-tyrosine, Fmoc-Tyr(tBu)-OH

F0506-1G F0506-25G F0506-5G
TCF0506-1GEA 17 USD
TCF0506-1G TCF0506-25G TCF0506-5G
FMOC-O-tert-butyl-L-tyrosine ≥98.0% (by HPLC, titration analysis)
FMOC-O-tert-butyl-L-tyrosine

CAS Number: 71989-38-3
Molecular Formula: C28H29NO5
Molecular Weight: 459.54
Purity/Analysis Method: >98.0% (T,HPLC)
Form: Crystal
Melting point (°C): 153
Specific rotation [a]20/D: -30 deg (C=1, DMF)
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Peptide Synthesis, Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: O-tert-Butyl-Nalpha-Fmoc-L-tyrosine, Nalpha-Fmoc-O-tert-butyl-L-tyrosine, Fmoc-Tyr(tBu)-OH

Formula: C₂₈H₂₉NO₅
MW: 459.54 g/mol
Melting Pt: 153 °C
MDL Number: MFCD00037129
CAS Number: 71989-38-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f135c611120fb0097a312553058be9555c075cd3&vid=f135c611120fb0097a312553058be9555c075cd3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T,HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR